Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146323.1 | NP_001139795.1 | 3 - 4 | 531 | 131 | |
NM_001146324.1 | NP_001139796.1 | 3 - 4 | 927 | 131 | ||
NM_080634.4 | NP_542365.3 | 3 - 4 | 927 | 131 | ||
GenBank mRNA | AF393747.1 | - | 3 - 4 | 916 | 131 | |
AF393780.1 | - | 3 - 4 | 919 | 131 | ||
AK034377.1 | - | 3 - 4 | 898 | 131 | ||
AK036663.1 | - | 3 - 4 | 935 | 131 | ||
AK037829.1 | - | 3 - 4 | 426 | 131 | ||
AK049204.1 | - | 3 - 4 | 920 | 131 | ||
AK079896.1 | - | 3 - 4 | 919 | 131 | ||
AK086167.1 | - | 3 - 4 | 916 | 131 | ||
AK149359.1 | - | 3 - 4 | 913 | 131 | ||
AK155497.1 | - | 3 - 4 | 894 | 131 | ||
AK157953.1 | - | 3 - 4 | 912 | 131 | ||
AK166194.1 | - | 3 - 4 | 913 | 131 | ||
AK170581.1 | - | 3 - 4 | 443 | 131 | ||
AK171997.1 | - | 3 - 4 | 519 | 131 | ||
BC058235.1 | - | 3 - 4 | 901 | 6307400 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |