Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164370.1 | NP_001157842.1 | 1 - 2 | 193 | 72 | |
GenBank mRNA | AK020763.1 | - | 1 - 2 | 69 | 72 | |
AK046422.1 | - | 1 - 2 | 192 | 72 | ||
AK050030.1 | - | 1 - 2 | 179 | 72 | ||
AK077915.1 | - | 1 - 2 | 173 | 72 | ||
AK085510.1 | - | 1 - 2 | 188 | 72 | ||
AK163490.1 | - | 1 - 2 | 173 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |