Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100027096.mm.1 |
NM_001134741.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027096.mm.1 |
NM_001134741.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027096.mm.1 |
TC0100003316.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027098.mm.1 |
NM_001134741.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027098.mm.1 |
NM_001134741.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027098.mm.1 |
NM_001277730.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027098.mm.1 |
TC0100003316.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027094.mm.1 |
NM_001134741.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027094.mm.1 |
NM_001134741.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027094.mm.1 |
NM_001277730.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027094.mm.1 |
TC0100003316.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027101.mm.1 |
NM_001277730.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027101.mm.1 |
TC0100003316.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027099.mm.1 |
NM_001134741.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027099.mm.1 |
NM_001134741.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027099.mm.1 |
TC0100003316.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027097.mm.1 |
NM_001134741.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027097.mm.1 |
NM_001134741.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027097.mm.1 |
NM_001277730.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027097.mm.1 |
TC0100003316.mm.1 |
MTA TC |