Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026876.3 | NP_081152.2 | 13 - 14 | 2034 | 92 | |
GenBank mRNA | AK004493.1 | - | 5 - 6 | 527 | 92 | |
BC022682.1 | - | 8 - 9 | 1070 | 92 | ||
BC037478.1 | - | 9 - 10 | 1214 | 92 | ||
BC064024.1 | - | 13 - 14 | 1832 | 92 | ||
BC110425.1 | - | 13 - 14 | 2017 | 5369595 | 92 | |
BC132299.1 | - | 13 - 14 | 1892 | 40130576 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |