Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC096468.1 | - | 1 - 2 | 449 | 3665952 | 69 |
BC121828.1 | - | 1 - 2 | 464 | 40107316 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |