Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198415.2 | NP_940807.1 | 8 - 9 | 1098 | 71 | |
GenBank mRNA | BC061221.1 | - | 8 - 9 | 1098 | 30296538 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |