Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173392.4 | NP_775568.1 | 13 - 14 | 4061 | 83 | |
GenBank mRNA | AK080779.1 | - | 9 - 10 | 1870 | 83 | |
AK122241.1 | - | 13 - 14 | 4042 | 83 | ||
BC042669.1 | - | 13 - 14 | 4059 | 4235024 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |