Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177129.5 | NP_796103.2 | 7 - 8 | 1104 | 60 | |
GenBank mRNA | AK147573.1 | - | 6 - 7 | 1232 | 60 | |
AK147602.1 | - | 7 - 8 | 1105 | 60 | ||
AK147658.1 | - | 6 - 7 | 1232 | 60 | ||
AK147673.1 | - | 6 - 7 | 1257 | 60 | ||
BC066106.1 | - | 7 - 8 | 1024 | 30362931 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |