Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271884.1 | NP_001258813.1 | 20 - 21 | 2309 | 59 | |
NM_207031.2 | NP_996914.1 | 20 - 21 | 2309 | 59 | ||
GenBank mRNA | BC116706.1 | - | 20 - 21 | 2275 | 40096570 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |