Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111312.1 | NP_001104782.1 | 4 - 5 | 416 | 88 | |
NM_008515.4 | NP_032541.1 | 12 - 13 | 973 | 88 | ||
GenBank mRNA | AF045573.1 | - | 12 - 13 | 910 | 88 | |
AK041162.1 | - | 4 - 5 | 367 | 88 | ||
BC144956.1 | - | 14 - 15 | 931 | 9053504 | 88 | |
BC145642.1 | - | 14 - 15 | 931 | 40130945 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |