Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113188.1 | NP_001106659.1 | 6 - 7 | 551 | 127 | |
NM_001113189.1 | NP_001106660.1 | 6 - 7 | 551 | 127 | ||
NM_008053.2 | NP_032079.1 | 6 - 7 | 551 | 127 | ||
GenBank mRNA | BC019139.1 | - | 6 - 7 | 532 | 5041635 | 127 |
X90875.1 | - | 6 - 7 | 530 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |