Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025456.3 | NP_079732.1 | 2 - 3 | 336 | 66 | |
GenBank mRNA | AK007442.1 | - | 2 - 3 | 291 | 66 | |
AK012800.1 | - | 2 - 3 | 293 | 66 | ||
AK145027.1 | - | 2 - 3 | 66 | |||
AK145041.1 | - | 2 - 3 | 66 | |||
AY882557.1 | - | 2 - 3 | 255 | 66 | ||
BC037622.1 | - | 2 - 3 | 363 | 5373275 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |