Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300012997.mm.1 |
NM_001195031.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012997.mm.1 |
NM_001195031.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012997.mm.1 |
TC0300001631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013002.mm.1 |
NM_001195031.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013002.mm.1 |
NM_001195031.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013002.mm.1 |
NM_053182.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013002.mm.1 |
NM_053182.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013002.mm.1 |
TC0300001631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013001.mm.1 |
NM_001195031.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013001.mm.1 |
NM_001195031.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013001.mm.1 |
NM_053182.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013001.mm.1 |
NM_053182.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013001.mm.1 |
TC0300001631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012999.mm.1 |
NM_001195031.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012999.mm.1 |
NM_001195031.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012999.mm.1 |
NM_053182.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012999.mm.1 |
NM_053182.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012999.mm.1 |
TC0300001631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300013000.mm.1 |
NM_001195031.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013000.mm.1 |
NM_001195031.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013000.mm.1 |
NM_053182.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013000.mm.1 |
NM_053182.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300013000.mm.1 |
TC0300001631.mm.1 |
MTA TC |