Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025975.5 | NP_080251.3 | 3 - 4 | 284 | 115 | |
GenBank mRNA | AK010186.1 | - | 3 - 4 | 218 | 115 | |
AK088277.1 | - | 3 - 4 | 285 | 115 | ||
BC030032.1 | - | 3 - 4 | 211 | 1332100 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |