Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199349.1 | NP_001186278.1 | 3 - 4 | 456 | 87 | |
GenBank mRNA | AK031264.1 | - | 3 - 4 | 213 | 87 | |
AK054364.1 | - | 3 - 4 | 256 | 87 | ||
AK080225.1 | - | 3 - 4 | 212 | 87 | ||
BY727540.1 | - | 3 - 4 | 457 | 87 | ||
EF516990.1 | - | 3 - 4 | 214 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |