Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162415.1 | NP_001155887.1 | 9 - 10 | 864 | 98 | |
NM_001162416.1 | NP_001155888.1 | 7 - 8 | 657 | 98 | ||
NM_008825.4 | NP_032851.2 | 8 - 9 | 758 | 98 | ||
NR_027859.1 | 8 - 9 | 758 | 98 | |||
GenBank mRNA | AK029659.1 | - | 9 - 10 | 865 | 98 | |
AK146744.1 | - | 8 - 9 | 752 | 98 | ||
BC018418.1 | - | 8 - 9 | 715 | 3979400 | 98 | |
BC051014.1 | - | 7 - 8 | 671 | 6493944 | 98 | |
EU616667.1 | - | 7 - 8 | 656 | 98 | ||
EU616668.1 | - | 7 - 8 | 656 | 98 | ||
EU616669.1 | - | 7 - 8 | 656 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |