Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019411.4 | NP_062284.1 | 4 - 5 | 805 | 63 | |
GenBank mRNA | AF076192.1 | - | 4 - 5 | 775 | 63 | |
AK076110.1 | - | 4 - 5 | 806 | 63 | ||
AK172644.1 | - | 4 - 5 | 778 | 63 | ||
AK195883.1 | - | 2 - 3 | 270 | 63 | ||
BC003856.1 | - | 4 - 5 | 782 | 3484084 | 63 | |
BC054458.1 | - | 4 - 5 | 765 | 6493866 | 63 | |
Z67745.1 | - | 4 - 5 | 580 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |