Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290741.1 | NP_001277670.1 | 5 - 6 | 563 | 116 | |
NM_212484.2 | NP_997649.1 | 6 - 7 | 917 | 116 | ||
GenBank mRNA | BC049984.1 | - | 1 - 2 | 87 | 116 | |
BC062950.1 | - | 6 - 7 | 905 | 6840162 | 116 | |
CF746424.1 | - | 4 - 5 | 397 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |