Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145926.3 | NP_666038.3 | 5 - 6 | 911 | 61 | |
GenBank mRNA | AB053218.1 | - | 5 - 6 | 613 | 61 | |
AK155336.1 | - | 5 - 6 | 912 | 61 | ||
AK181196.1 | - | 2 - 3 | 89 | 61 | ||
CX205743.1 | - | 2 - 3 | 171 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |