Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178247.3 | NP_839978.1 | 2 - 3 | 134 | 96 | |
GenBank mRNA | AK145642.1 | - | 2 - 3 | 135 | 96 | |
AK166752.1 | - | 2 - 3 | 135 | 96 | ||
BC115806.1 | - | 1 - 2 | 75 | 40106955 | 96 | |
BC115901.1 | - | 1 - 2 | 75 | 40106956 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |