Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF128214.1 | - | 6 - 7 | 899 | 87 | |
AF128215.1 | - | 6 - 7 | 899 | 87 | ||
AK162981.1 | - | 6 - 7 | 923 | 87 | ||
BC103608.1 | - | 6 - 7 | 905 | 40046200 | 87 | |
BC103609.1 | - | 6 - 7 | 905 | 40046202 | 87 | |
BC103610.1 | - | 6 - 7 | 905 | 40046204 | 87 | |
BC103614.1 | - | 6 - 7 | 905 | 40046206 | 87 | |
M86671.1 | - | 6 - 7 | 900 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |