Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024200.4 | NP_077162.2 | 4 - 5 | 628 | 84 | |
GenBank mRNA | AK018181.1 | - | 4 - 5 | 84 | ||
AK046313.1 | - | 4 - 5 | 581 | 84 | ||
AK141611.1 | - | 4 - 5 | 629 | 84 | ||
AK220169.1 | - | 4 - 5 | 616 | 84 | ||
AY174062.1 | - | 3 - 4 | 757 | 84 | ||
BC056641.1 | - | 4 - 5 | 587 | 5097345 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |