Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089100.1 | - | 2 - 3 | 206 | 80 | |
BC080774.1 | - | 2 - 3 | 174 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |