Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007628.3 | NP_031654.2 | 3 - 4 | 522 | 109 | |
GenBank mRNA | AK077114.1 | - | 3 - 4 | 528 | 109 | |
BC120518.1 | - | 2 - 3 | 437 | 40129441 | 109 | |
BC125436.1 | - | 2 - 3 | 437 | 40129951 | 109 | |
X84311.1 | - | 3 - 4 | 521 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |