Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005509.2 | NP_001005509.1 | 2 - 3 | 334 | 93 | |
GenBank mRNA | AK050971.1 | - | 2 - 3 | 113 | 93 | |
AK078519.1 | - | 2 - 3 | 113 | 93 | ||
AK145275.1 | - | 2 - 3 | 290 | 93 | ||
AK161904.1 | - | 2 - 3 | 115 | 93 | ||
BC030447.1 | - | 2 - 3 | 122 | 5400637 | 93 | |
BC082557.1 | - | 2 - 3 | 139 | 30606936 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |