Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172862.3 | NP_766450.2 | 1 - 2 | 5410 | 82 | |
GenBank mRNA | AB236662.1 | - | 1 - 2 | 5410 | 82 | |
AJ833643.1 | - | 1 - 2 | 5139 | 82 | ||
CN536459.1 | - | 1 - 2 | 75 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mus musculus 1700 Probe spans exons |