Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133198.2 | NP_573461.2 | 15 - 16 | 1922 | 90 | |
GenBank mRNA | AF288783.1 | - | 15 - 16 | 1864 | 90 | |
AK140321.1 | - | 15 - 16 | 1869 | 90 | ||
AK145989.1 | - | 15 - 16 | 1923 | 90 | ||
AK149491.1 | - | 2097 | 90 | |||
AK167337.1 | - | 15 - 16 | 1869 | 90 | ||
BC013636.1 | - | 15 - 16 | 1856 | 4196237 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |