Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010712.2 | NP_034842.2 | 2 - 3 | 399 | 65 | |
GenBank mRNA | AB221565.1 | - | 1 - 2 | 180 | 65 | |
AF135415.1 | - | 1 - 2 | 184 | 65 | ||
BC049834.1 | - | 2 - 3 | 399 | 6493203 | 65 | |
U89343.1 | - | 1 - 2 | 162 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |