Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008856.3 | NP_032882.2 | 5 - 6 | 822 | 69 | |
GenBank mRNA | AK076352.1 | - | 5 - 6 | 1021 | 69 | |
AK145817.1 | - | 5 - 6 | 1068 | 69 | ||
AK155951.1 | - | 6 - 7 | 1165 | 69 | ||
AK164044.1 | - | 5 - 6 | 1021 | 69 | ||
BC031121.1 | - | 5 - 6 | 822 | 5059633 | 69 | |
D90242.1 | - | 5 - 6 | 759 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |