Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130174.2 | NP_001123646.1 | 14 - 15 | 952 | 67 | |
NM_001130175.2 | NP_001123647.4 | 14 - 15 | 948 | 67 | ||
NM_001130176.1 | NP_001123648.1 | 13 - 14 | 848 | 67 | ||
NM_001130177.2 | NP_001123649.1 | 13 - 14 | 940 | 67 | ||
NM_001130178.2 | NP_001123650.1 | 14 - 15 | 931 | 67 | ||
NM_001130179.2 | NP_001123651.4 | 14 - 15 | 927 | 67 | ||
NM_001130180.2 | NP_001123652.1 | 13 - 14 | 919 | 67 | ||
NM_001130181.2 | NP_001123653.4 | 13 - 14 | 915 | 67 | ||
NM_011619.3 | NP_035749.1 | 12 - 13 | 910 | 67 | ||
GenBank mRNA | AK146945.1 | - | 14 - 15 | 897 | 67 | |
AK168393.1 | - | 13 - 14 | 885 | 67 | ||
AK169061.1 | - | 14 - 15 | 877 | 67 | ||
BC063753.1 | - | 14 - 15 | 928 | 30293959 | 67 | |
L47549.1 | - | 14 - 15 | 845 | 67 | ||
L47550.1 | - | 14 - 15 | 830 | 67 | ||
L47551.1 | - | 13 - 14 | 831 | 67 | ||
L47552.1 | - | 13 - 14 | 840 | 67 | ||
L47553.1 | - | 13 - 14 | 847 | 67 | ||
L47599.1 | - | 14 - 15 | 851 | 67 | ||
L47600.1 | - | 12 - 13 | 839 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |