Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111311.1 | NP_001104781.1 | 6 - 7 | 786 | 72 | |
NM_001111312.1 | NP_001104782.1 | 7 - 8 | 745 | 72 | ||
GenBank mRNA | AK041162.1 | - | 7 - 8 | 696 | 72 | |
AK077637.1 | - | 6 - 7 | 791 | 72 | ||
AK133661.1 | - | 6 - 7 | 767 | 72 | ||
AK134120.1 | - | 6 - 7 | 778 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |