Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011137.3 | NP_035267.2 | 12 - 13 | 1477 | 83 | |
NM_198932.3 | NP_945150.2 | 12 - 13 | 1477 | 83 | ||
NM_198933.3 | NP_945151.2 | 12 - 13 | 1549 | 83 | ||
NM_198934.3 | NP_945152.2 | 12 - 13 | 1549 | 83 | ||
GenBank mRNA | AF508939.1 | - | 12 - 13 | 1458 | 83 | |
AJ489474.1 | - | 12 - 13 | 1549 | 83 | ||
AJ489475.1 | - | 11 - 12 | 1366 | 83 | ||
AK028237.1 | - | 12 - 13 | 1479 | 83 | ||
AY177625.1 | - | 12 - 13 | 1458 | 83 | ||
BC066201.1 | - | 12 - 13 | 1454 | 83 | ||
S65461.1 | - | 11 - 12 | 1377 | 83 | ||
X56230.1 | - | 11 - 12 | 1554 | 83 | ||
X68362.1 | - | 12 - 13 | 1450 | 83 | ||
X68363.1 | - | 12 - 13 | 1448 | 83 | ||
X68364.1 | - | 12 - 13 | 1464 | 83 | ||
X70324.1 | - | 10 - 11 | 1309 | 83 | ||
X70325.1 | - | 2 - 3 | 290 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |