Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025388.2 | NP_079664.1 | 4 - 5 | 387 | 79 | |
GenBank mRNA | AK003070.1 | - | 3 - 4 | 289 | 79 | |
AK003897.1 | - | 4 - 5 | 388 | 79 | ||
AK011260.1 | - | 3 - 4 | 278 | 79 | ||
BC021936.1 | - | 4 - 5 | 348 | 5124904 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |