Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286003.1 | NP_001272932.1 | 9 - 10 | 1455 | 117 | |
NM_028776.5 | NP_083052.1 | 9 - 10 | 1430 | 117 | ||
GenBank mRNA | AK004809.2 | - | 9 - 10 | 1170 | 117 | |
AK030824.1 | - | 9 - 10 | 1405 | 117 | ||
AK044100.1 | - | 9 - 10 | 1416 | 117 | ||
BC043085.1 | - | 9 - 10 | 1131 | 6417440 | 117 | |
BC066800.1 | - | 9 - 10 | 1115 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |