Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pm20d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178079.3 | NP_835180.2 | 10 - 11 | 1147 | 61 | |
GenBank mRNA | AK028883.1 | - | 10 - 11 | 1144 | 61 | |
AK128969.1 | - | 10 - 11 | 1153 | 61 | ||
BC025830.1 | - | 7 - 8 | 637 | 61 | ||
BC049922.1 | - | 3 - 4 | 184 | 61 | ||
BC120725.1 | - | 10 - 11 | 1147 | 40129648 | 61 | |
BC125395.1 | - | 10 - 11 | 1147 | 40129910 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |