Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco6c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028942.4 | NP_083218.1 | 12 - 13 | 2113 | 146 | |
GenBank mRNA | AK016647.1 | - | 12 - 13 | 2061 | 146 | |
AK029505.1 | - | 12 - 13 | 2114 | 146 | ||
AK132668.1 | - | 12 - 13 | 2064 | 146 | ||
AK161249.1 | - | 12 - 13 | 2064 | 146 | ||
BC139066.1 | - | 12 - 13 | 2016 | 8862088 | 146 | |
BC139067.1 | - | 12 - 13 | 2016 | 8862089 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |