Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172157.1 | NP_001165628.1 | 7 - 8 | 802 | 115 | |
NM_153530.2 | NP_705758.1 | 6 - 7 | 760 | 115 | ||
GenBank mRNA | AK029652.1 | - | 7 - 8 | 816 | 115 | |
AK031180.1 | - | 7 - 8 | 785 | 115 | ||
AK040613.1 | - | 8 - 9 | 857 | 115 | ||
BC036177.1 | - | 6 - 7 | 721 | 5004341 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |