Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172157.1 | NP_001165628.1 | 6 - 7 | 574 | 115 | |
GenBank mRNA | AK031180.1 | - | 6 - 7 | 557 | 115 | |
AK184309.1 | - | 2 - 3 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |