Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011495.2 | NP_035625.2 | 2 - 3 | 419 | 72 | |
NM_173169.2 | NP_775261.2 | 2 - 3 | 419 | 72 | ||
NR_045576.1 | 2 - 3 | 419 | 72 | |||
GenBank mRNA | AK006827.1 | - | 2 - 3 | 325 | 72 | |
AK045082.1 | - | 2 - 3 | 343 | 72 | ||
AK137080.1 | - | 2 - 3 | 331 | 72 | ||
AK137471.1 | - | 2 - 3 | 330 | 72 | ||
AK164465.1 | - | 2 - 3 | 329 | 72 | ||
BC026785.1 | - | 2 - 3 | 329 | 5150173 | 72 | |
BC057940.1 | - | 2 - 3 | 316 | 5323295 | 72 | |
L29479.1 | - | 2 - 3 | 321 | 72 | ||
L29480.1 | - | 2 - 3 | 321 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |