Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284524.1 | NP_001271453.1 | 8 - 9 | 955 | 73 | |
NM_001284525.1 | NP_001271454.1 | 8 - 9 | 991 | 73 | ||
NM_027979.2 | NP_082255.1 | 8 - 9 | 955 | 73 | ||
GenBank mRNA | AK009012.1 | - | 8 - 9 | 956 | 73 | |
AY458654.1 | - | 8 - 9 | 955 | 73 | ||
AY536287.1 | - | 7 - 8 | 829 | 73 | ||
BC138765.1 | - | 8 - 9 | 957 | 8861787 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |