Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009791.4 | NP_033921.3 | 8 - 9 | 2689 | 71 | |
GenBank mRNA | AF533752.1 | - | 8 - 9 | 2667 | 71 | |
AK052326.1 | - | 8 - 9 | 2690 | 71 | ||
AY971958.1 | - | 8 - 9 | 2530 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |