Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253708.1 | NP_001240637.1 | 7 - 8 | 2036 | 76 | |
NM_001253710.1 | NP_001240639.1 | 6 - 7 | 987 | 76 | ||
GenBank mRNA | AF231110.1 | - | 6 - 7 | 1476 | 76 | |
BC096600.1 | - | 6 - 7 | 948 | 4976244 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |