Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm31063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK081945.1 | - | 341 | 138 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MIS18 binding protein 1 | Mis18bp1 |
6720403H11, C79407, Knl2, mKIAA1903 |
-15.3kb | Sense | Chr. 12 - 65132734 - 65172580 on NCBI Build GRCm38 |
Downstream |
WD repeat domain 20, retrogene |
Wdr20rt |
4921538B03Rik, 4930427E19Rik, Wdr20, Wdr20b |
28.6kb | Anti-sense | Chr. 12 - 65225517 - 65228454 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons |