Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028399.1 | NP_082675.1 | 3 - 4 | 428 | 109 | |
GenBank mRNA | AK013634.1 | - | 3 - 4 | 375 | 109 | |
AK036772.1 | - | 376 | 109 | |||
AK049863.1 | - | 3 - 4 | 373 | 109 | ||
AK152399.1 | - | 3 - 4 | 376 | 109 | ||
AK154439.1 | - | 3 - 4 | 386 | 109 | ||
AK156231.1 | - | 397 | 109 | |||
BC054122.1 | - | 3 - 4 | 428 | 6509024 | 109 | |
JX965957.1 | - | 3 - 4 | 364 | 109 | ||
JX965958.1 | - | 3 - 4 | 364 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |