Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160307.1 | NP_001153779.1 | 4 - 5 | 415 | 97 | |
NM_198028.3 | NP_932145.3 | 5 - 6 | 591 | 97 | ||
GenBank mRNA | AK036537.1 | - | 5 - 6 | 569 | 97 | |
AK052580.1 | - | 4 - 5 | 416 | 97 | ||
BC029736.1 | - | 5 - 6 | 532 | 3676050 | 97 | |
BC069938.1 | - | 5 - 6 | 551 | 5035202 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |