Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ispd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK045940.1 | - | 9 - 10 | 1561 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |