Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133198.2 | NP_573461.2 | 1 - 2 | 342 | 72 | |
GenBank mRNA | AF288783.1 | - | 1 - 2 | 284 | 72 | |
AK041189.1 | - | 1 - 2 | 289 | 72 | ||
AK140321.1 | - | 1 - 2 | 289 | 72 | ||
AK145989.1 | - | 1 - 2 | 343 | 72 | ||
AK167337.1 | - | 1 - 2 | 289 | 72 | ||
BC013636.1 | - | 1 - 2 | 276 | 4196237 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |