Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133239.2 | NP_573502.2 | 9 - 10 | 3910 | 76 | |
GenBank mRNA | AF406641.1 | - | 9 - 10 | 3875 | 76 | |
AF492496.1 | - | 8 - 9 | 3751 | 76 | ||
AK164413.1 | - | 1 - 2 | 874 | 76 | ||
BC031378.1 | - | 2 - 3 | 970 | 76 | ||
BC053331.1 | - | 4 - 5 | 2153 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |