Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130456.3 | NP_569723.1 | 5 - 6 | 796 | 81 | |
GenBank mRNA | AJ302048.1 | - | 5 - 6 | 743 | 81 | |
AK052746.1 | - | 5 - 6 | 805 | 81 | ||
AY050309.1 | - | 5 - 6 | 819 | 81 | ||
BC067401.1 | - | 5 - 6 | 796 | 30470258 | 81 | |
FJ715770.1 | - | 5 - 6 | 804 | 81 | ||
FJ715771.1 | - | 5 - 6 | 834 | 81 | ||
FJ715772.1 | - | 5 - 6 | 834 | 81 | ||
FJ715773.1 | - | 5 - 6 | 834 | 81 | ||
FJ715775.1 | - | 5 - 6 | 834 | 81 | ||
FJ715776.1 | - | 5 - 6 | 834 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |