Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007753.2 | NP_031779.1 | 5 - 6 | 478 | 122 | |
GenBank mRNA | AK088906.1 | - | 5 - 6 | 479 | 122 | |
BC108409.1 | - | 5 - 6 | 467 | 122 | ||
J05118.1 | - | 5 - 6 | 474 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |